Learn More
TARGETMOL CHEMICALS INC Gypsogenin-3-O-glucuronide 5MG
SDP
Catalog No. 502876656
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 1. Gypsogenin-3-O-glucuronide (Methyl gypsogenin 3-O-beta-D-glucuronopy) shows inhibitory activity toward pancreatic lipase with IC5 value of .29 mM, and the free carboxylic acid groups in position 28 within its chemical structures are required for enhancement of pancreatic lipase inhibition. Purity 99.33%
Catalog No. 50-287-6656 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S13125MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 1. Gypsogenin-3-O-glucuronide (Methyl gypsogenin 3-O-beta-D-glucuronopy) shows inhibitory activity toward pancreatic lipase with IC5 value of .29 mM, and the free carboxylic acid groups in position 28 within its chemical structures are required for enhancement of pancreatic lipase inhibition. Purity 99.33%