Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gypsogenin | 639-14-5 | 99.5% | 25 MG
SDP

Catalog No. 5000231204
Encompass_Preferred

Gypsogenin is a pentacyclic triterpenoid sapogenin (CAS 639-14-5) used for research. It has molecular formula C30H46O4 and a molecular weight of 470.68 g/mol, and is reported to show antiangiogenic activity and cytotoxicity against H460 cells. Typically supplied as a 25 mg research sample.

  • High purity (≈99.5%).
  • Soluble in DMSO (25 mg/mL); may require sonication and freshly opened DMSO.
  • Recommended storage: 4°C; in solvent, -80°C (6 months) or -20°C (1 month); protect from light.
  • Intended for research use; not for human or clinical use.

Catalog No. 50-002-31204 Supplier Medchemexpress LLC Supplier No. HY12138225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gypsogenin is a pentacyclic triterpenoid sapogenin (CAS 639-14-5) used for research. It has molecular formula C30H46O4 and a molecular weight of 470.68 g/mol, and is reported to show antiangiogenic activity and cytotoxicity against H460 cells. Typically supplied as a 25 mg research sample.

  • High purity (≈99.5%).
  • Soluble in DMSO (25 mg/mL); may require sonication and freshly opened DMSO.
  • Recommended storage: 4°C; in solvent, -80°C (6 months) or -20°C (1 month); protect from light.
  • Intended for research use; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.