Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gypsogenin | 639-14-5 | 99.5% | 5 MG
SDP

Catalog No. 5000232086
Encompass_Preferred

Gypsogenin is an oleanane-type triterpenoid shown to have antiangiogenic activity and significant cytotoxicity against H460 cells. It is supplied as a solid research reagent, typically dissolved in DMSO for biological assays and stored protected from light.

  • Natural triterpenoid with antiangiogenic activity.
  • Shows significant cytotoxicity against H460 cell line.
  • Empirical formula C30H46O4 and molecular weight 470.7 g/mol.
  • Purity typically 99.5% as supplied.
  • Soluble in DMSO at 25 mg/mL; ultrasonic assistance recommended.
  • Storage: 4°C protected from light; in solvent store at -80°C up to 6 months.
  • Supplied as a 5 mg solid suitable for laboratory research use.

Catalog No. 50-002-32086 Supplier Medchemexpress LLC Supplier No. HY1213825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gypsogenin is an oleanane-type triterpenoid shown to have antiangiogenic activity and significant cytotoxicity against H460 cells. It is supplied as a solid research reagent, typically dissolved in DMSO for biological assays and stored protected from light.

  • Natural triterpenoid with antiangiogenic activity.
  • Shows significant cytotoxicity against H460 cell line.
  • Empirical formula C30H46O4 and molecular weight 470.7 g/mol.
  • Purity typically 99.5% as supplied.
  • Soluble in DMSO at 25 mg/mL; ultrasonic assistance recommended.
  • Storage: 4°C protected from light; in solvent store at -80°C up to 6 months.
  • Supplied as a 5 mg solid suitable for laboratory research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.