Learn More
AMPLYUS LLC miniPCR bio Gyro™ Microcentrifuge, fixed speed
SDP
Catalog No. NC1661598
Encompass_Preferred
Gyro™ microcentrifuge, fixed speed, for bead-based purification of nucleic acids and proteins. Single speed: 10,000 rpm. Holds 0.2, 0.5, 1.5, and 2.0 ml tubes.
Catalog No. NC1661598 Supplier AMPLYUS LLC Supplier No. QP180001
Gyro™ microcentrifuge, fixed speed, for bead-based purification of nucleic acids and proteins. Single speed: 10,000 rpm. Holds 0.2, 0.5, 1.5, and 2.0 ml tubes.