Learn More
Braintree Scientific Gyro Mini Nutating Mixer
SDP
Catalog No. 501955829
Encompass
GyroMini is a smaller version of the popular GyroTwister. Its small size makes it ideal for mixing samples in blood and centrifuge tubes. GyroMini combines motions of an orbital shaker and a rocker to produce a three dimensional action. The result is a thorough yet gentle mixing without foaming. Both speed and tilt angle of this small shaker are fixed to provide the optimal level of agitation for mixing samples in small containers. GyroMini is supplied with a dimpled mat for holding tubes firmly in place. For temperature sensitive samples, the GyroMini can be used in a cold room. Specifications: Speed range: 24 rpm; Motion/ Pitch: 3-D/fixed 20; Maximum load: 1.75 lb/.8 kg; Ambient operating range: +4 to 65C; Dimensions (W x D x H): 9.75" x 7.5" x 7"/24.8cm x 19.1cm x 17.8cm. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5829 Supplier Braintree Scientific Supplier No. S0500120V
GyroMini is a smaller version of the popular GyroTwister. Its small size makes it ideal for mixing samples in blood and centrifuge tubes. GyroMini combines motions of an orbital shaker and a rocker to produce a three dimensional action. The result is a thorough yet gentle mixing without foaming. Both speed and tilt angle of this small shaker are fixed to provide the optimal level of agitation for mixing samples in small containers. GyroMini is supplied with a dimpled mat for holding tubes firmly in place. For temperature sensitive samples, the GyroMini can be used in a cold room. Specifications: Speed range: 24 rpm; Motion/ Pitch: 3-D/fixed 20; Maximum load: 1.75 lb/.8 kg; Ambient operating range: +4 to 65C; Dimensions (W x D x H): 9.75" x 7.5" x 7"/24.8cm x 19.1cm x 17.8cm. **Sold for research purposes only and not for human use**