Learn More
Gyros Inc Gyrolab Bioaffy 200
SDP
Catalog No. NC9914524
Encompass
1 Gyrolab Bioaffy CD microlaboratory, 200 nL sample volume, 112 microstructures.
Catalog No. NC9914524 Supplier Gyros Inc Supplier No. P0004180
1 Gyrolab Bioaffy CD microlaboratory, 200 nL sample volume, 112 microstructures.