Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gyros Inc Gyrolab Generic PK Kit, for quantifying human therapeutic antibodies, ready-to-use, includes pre-labeled reagents, 96 data points, analytical range 20 ng/mL to 14 μg/mL
SDP

Catalog No. NC1463677
Encompass

The Gyrolab Generic PK Kit is a ready-to-use immunoassay kit designed for the accurate and robust quantification of human therapeutic antibodies in non-human matrices during pre-clinical pharmacokinetic (PK) studies. This kit efficiently quantifies monoclonal antibodies (mAbs) of IgG1, IgG2, and IgG4 subtypes in various animal matrices, offering a streamlined workflow for early-stage biotherapeutic development.

  • Application: Pre-clinical PK studies
  • Compatible matrices: Mouse, rat, cynomolgus monkey, rabbit, dog
  • Data points per run: 96
  • Run time: 70 min
  • Analytical range: 20 ng/mL to 14 μg/mL
  • Included CD type: Gyrolab Bioaffy 1000 HC CD
  • Sample dilution buffers: Type E and Type F included
  • Kit format: Ready-to-use
  • Selectivity: Broad for intact human IgG
  • Standard included: No
  • Composition: Pre-labeled capture and detection reagents, buffers, consumables, 1 CD

Catalog No. NC1463677 Supplier Gyros Inc Supplier No. P0020499
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Gyrolab Generic PK Kit is a ready-to-use immunoassay kit designed for the accurate and robust quantification of human therapeutic antibodies in non-human matrices during pre-clinical pharmacokinetic (PK) studies. This kit efficiently quantifies monoclonal antibodies (mAbs) of IgG1, IgG2, and IgG4 subtypes in various animal matrices, offering a streamlined workflow for early-stage biotherapeutic development.

  • Application: Pre-clinical PK studies
  • Compatible matrices: Mouse, rat, cynomolgus monkey, rabbit, dog
  • Data points per run: 96
  • Run time: 70 min
  • Analytical range: 20 ng/mL to 14 μg/mL
  • Included CD type: Gyrolab Bioaffy 1000 HC CD
  • Sample dilution buffers: Type E and Type F included
  • Kit format: Ready-to-use
  • Selectivity: Broad for intact human IgG
  • Standard included: No
  • Composition: Pre-labeled capture and detection reagents, buffers, consumables, 1 CD

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.