Learn More
Gyros Inc Gyrolab Wash Buffer pH 11, 10/pk
SDP
Catalog No. NC0575330
Encompass
Biodegradable high pH washing buffer. Package of 10x10 grams. 10 grams is required for 1 liter of needle wash buffer.
Catalog No. NC0575330 Supplier Gyros Inc Supplier No. P0020096
Biodegradable high pH washing buffer. Package of 10x10 grams. 10 grams is required for 1 liter of needle wash buffer.