Learn More
Abcam GYY 4137, H2S donor, 25MG
Catalog No. 502706313
Encompass_Preferred
MW 376.5 Da, Purity >95%. Water-soluble, slow-releasing hydrogen sulfide donor. Inhibits LPS-induced release of proinflammatory intermediates and increases synthesis of IL-10. Exhibits novel anti-cancer effects. Vasodilator and antihypertensive activity. Active *in vitro* and *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6313 Supplier Abcam Supplier No. AB14214525MG
MW 376.5 Da, Purity >95%. Water-soluble, slow-releasing hydrogen sulfide donor. Inhibits LPS-induced release of proinflammatory intermediates and increases synthesis of IL-10. Exhibits novel anti-cancer effects. Vasodilator and antihypertensive activity. Active *in vitro* and *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement