Learn More
Apexbio Technology LLC GYY 4137 morpholine salt(Synonyms: GYY4137, Morpholin-4-ium 4-methoxyphenyl(morpholino)phosphinodithioate, GYY-4137), 50mg, CAS: 106740-09-4.
SDP
Catalog No. 5000060199
Encompass_Preferred
GYY 4137 morpholine salt (CAS 106740-09-4) is a slow-releasing hydrogen sulfide (H S) donor that exerts biological activity by modulating signaling pathways involved in vasodilation and blood pressure regulation In experimental studies this compound demonstrates the capacity to suppress cellular proliferation induce apoptosis and arrest cell cycle progression Due to these properties GYY 4137 morpholine salt is widely utilized in research investigating hypertension inflammation and cancer biology
Catalog No. 50-000-60199 Supplier Apexbio Technology LLC Supplier No. B745850
GYY 4137 morpholine salt (CAS 106740-09-4) is a slow-releasing hydrogen sulfide (H S) donor that exerts biological activity by modulating signaling pathways involved in vasodilation and blood pressure regulation In experimental studies this compound demonstrates the capacity to suppress cellular proliferation induce apoptosis and arrest cell cycle progression Due to these properties GYY 4137 morpholine salt is widely utilized in research investigating hypertension inflammation and cancer biology