Learn More
Selleck Chemical LLC GYY4137-10mg
SDP
Supplier:  Selleck Chemical LLC S992010MG
Encompass_Preferred
GYY4137 is a novel, water-soluble hydrogen sulfide (H2S)–releasing molecule with vasodilator and antihypertensive activity. GYY4137 shows potent anti-hepatocellular carcinoma activity through blocking the STAT3 pathway. GYY4137 also shows anti-inflammatory activity.
Catalog No. 50-238-9907