Learn More
Apexbio Technology LLC GZD824, 25mg. Cas: 1421783-64-3 MFCD: MFCD26142656. Bcr-Abl inhibitor,novel orally bioavailable
SDP
Supplier:  Apexbio Technology LLC B140225
Encompass_Preferred
GZD824 is an orally bioavailable inhibitor of Bcr-Abl with IC50 values of 0.34 and 0.68 nM for Bcr-AblWT and Bcr-AblT315I, respectively [1]. Bcr-Abl is a fused protein that interacts with the interleukin-3 receptor(c) subunit and has tyrosine kinase activity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5228
 Restricted Item