Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GZD856 formic | 2804039-78-7 | 95.4% | 578.58 g/mol | C30H29F3N6O3 | 1 ML
SDP

Catalog No. 5000247130
Encompass_Preferred

GZD856 formic is a research-grade small-molecule inhibitor supplied as a 10 mM solution in DMSO for laboratory use. It is provided as the formic salt in a 1 mL vial and is intended for in vitro and exploratory studies needing a pre-dissolved, characterized compound.

  • Pre-formulated 10 mM solution for immediate use.
  • Supplied in a 1 mL vial for small-scale experiments.
  • Formic salt form to enhance solubility and stability.
  • Purity determined by HPLC, reported at 95.36%.
  • Molecular weight 578.58 g/mol; formula C30H29F3N6O3.

Catalog No. 50-002-47130 Supplier Medchemexpress LLC Supplier No. HY101489A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GZD856 formic is a research-grade small-molecule inhibitor supplied as a 10 mM solution in DMSO for laboratory use. It is provided as the formic salt in a 1 mL vial and is intended for in vitro and exploratory studies needing a pre-dissolved, characterized compound.

  • Pre-formulated 10 mM solution for immediate use.
  • Supplied in a 1 mL vial for small-scale experiments.
  • Formic salt form to enhance solubility and stability.
  • Purity determined by HPLC, reported at 95.36%.
  • Molecular weight 578.58 g/mol; formula C30H29F3N6O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.