Learn More
Aviva Systems Biology Corp GZMA Antibody - C-terminal region (ARP54794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GZMA. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GZMA. Alternate names: CTLA3, HFSPApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rat
Catalog No. 50-285-968