Learn More
Selleck Chemical LLC H-1152 dihydrochloride-5mg
SDP
Catalog No. 502390018
Encompass_Preferred
H-1152 dihydrochloride (2HCl) is a membrane-permeable and selective inhibitor of Rho-associated protein kinase (ROCK). H-1152 inhibits ROCK2, PKA, PKC, PKG, AuroraA and CaMK2 with IC50 of 0.0120 μM, 3.03 μM, 5.68 μM, 0.360 μM, 0.745 μM and 0.180 μM, respectively.
Catalog No. 50-239-0018 Supplier Selleck Chemical LLC Supplier No. S62145MG
H-1152 dihydrochloride (2HCl) is a membrane-permeable and selective inhibitor of Rho-associated protein kinase (ROCK). H-1152 inhibits ROCK2, PKA, PKC, PKG, AuroraA and CaMK2 with IC50 of 0.0120 μM, 3.03 μM, 5.68 μM, 0.360 μM, 0.745 μM and 0.180 μM, respectively.