Learn More
Selleck Chemical LLC H-1152 dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S621425MG
Encompass_Preferred
H-1152 dihydrochloride (2HCl) is a membrane-permeable and selective inhibitor of Rho-associated protein kinase (ROCK) H-1152 inhibits ROCK2 PKA PKC PKG AuroraA and CaMK2 with IC50 of 0 0120 M 3 03 M 5 68 M 0 360 M 0 745 M and 0 180 M respectively
Catalog No. 50-000-03774