Learn More
Medchemexpress LLC H-1152 dihydrochloride | 871543-07-6 | 99.1% | 392.34 g/mol | C16H23Cl2N3O2S | 10 MG
SDP
Catalog No. 5000287870
Encompass_Preferred
H-1152 dihydrochloride is a membrane-permeable, selective inhibitor of Rho-associated protein kinases (ROCK), reported with a Ki of 1.6 nM and an IC50 of 12 nM for ROCK2. Supplied as the dihydrochloride salt in solid form, it is intended for research use and offers formulation options for both in vitro and in vivo studies.
  • Potent inhibition of ROCK family kinases (Ki = 1.6 nM; IC50 ROCK2 = 12 nM).
  • Membrane-permeable for cellular assays.
  • High purity suitable for research applications.
  • Multiple solubility and formulation options for in vitro and in vivo use.
  • Available in small solid quantities convenient for laboratory experiments.
Catalog No. 50-002-87870 Supplier Medchemexpress LLC Supplier No. HY15720A10MG
H-1152 dihydrochloride is a membrane-permeable, selective inhibitor of Rho-associated protein kinases (ROCK), reported with a Ki of 1.6 nM and an IC50 of 12 nM for ROCK2. Supplied as the dihydrochloride salt in solid form, it is intended for research use and offers formulation options for both in vitro and in vivo studies.
  • Potent inhibition of ROCK family kinases (Ki = 1.6 nM; IC50 ROCK2 = 12 nM).
  • Membrane-permeable for cellular assays.
  • High purity suitable for research applications.
  • Multiple solubility and formulation options for in vitro and in vivo use.
  • Available in small solid quantities convenient for laboratory experiments.