Learn More
Medchemexpress LLC HY-112693 10mg Medchemexpress, H-151 CAS:941987-60-6 Purity:>98%
SDP
Catalog No. 501969143
Encompass_Preferred
Medchemexpress, HY-112693 10mg H-151 CAS:941987-60-6 H-151 is a highly potent, selective and covalent antagonist of STING, reduces TBK1 phosphorylation and suppresses human STING palmitoylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9143 Supplier Medchemexpress LLC Supplier No. HY11269310MG
Medchemexpress, HY-112693 10mg H-151 CAS:941987-60-6 H-151 is a highly potent, selective and covalent antagonist of STING, reduces TBK1 phosphorylation and suppresses human STING palmitoylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.