Learn More
Selleck Chemical LLC H-151
Supplier: Selleck Chemical LLC S665225MG
H-151 is a highly potent and covalent inhibitor of STING that has noteworthy inhibitory activity both in human cells and in vivo