Learn More
Selleck Chemical LLC H-151
SDP
Catalog No. 5000002365
Encompass_Preferred
H-151 is a highly potent and covalent inhibitor of STING that has noteworthy inhibitory activity both in human cells and in vivo
Catalog No. 50-000-02365 Supplier Selleck Chemical LLC Supplier No. S6652100MG
H-151 is a highly potent and covalent inhibitor of STING that has noteworthy inhibitory activity both in human cells and in vivo