Learn More
Medchemexpress LLC HY-112693 50mg , H-151 CAS:941987-60-6 Purity:>98%
SDP
Catalog No. 502259056
Encompass_Preferred
HY-112693 50mg H-151 CAS: 941987-60-6 H-151 is a potent, selective and covalent antagonist of STING that has noteworthy inhibitory activity both in cells and in vivo. H-151 reduces TBK1 phosphorylation and suppresses STING palmitoylation. H-151 can be used for the research of autoinflammatory disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9056 Supplier Medchemexpress LLC Supplier No. HY11269350MG
HY-112693 50mg H-151 CAS: 941987-60-6 H-151 is a potent, selective and covalent antagonist of STING that has noteworthy inhibitory activity both in cells and in vivo. H-151 reduces TBK1 phosphorylation and suppresses STING palmitoylation. H-151 can be used for the research of autoinflammatory disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.