Learn More
Selleck Chemical LLC H-151 5mg
SDP
Supplier:  Selleck Chemical LLC S66525MG
Encompass_Preferred
H-151 is a highly potent and covalent antagonist of STING that has noteworthy inhibitory activity both in human cells and in vivo. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3196