Learn More
Medchemexpress LLC H-151 Alkyne | 2244892-03-1 | 95.1% | 275.30 | 100 MG
SDP
Catalog No. 5000387108
Encompass_Preferred
H-151 Alkyne is a selective STING (Stimulator of interferon genes) inhibitor with potential for research in autoimmune diseases, including systemic lupus erythematosus, scleroderma, and autoinflammatory diseases.
  • Selective STING inhibitor
  • Promising for autoimmune disease research
  • Supplied as a solid
  • Light yellow to brown appearance
  • High purity of 95.08%
Catalog No. 50-003-87108 Supplier Medchemexpress LLC Supplier No. HY176566100MG
H-151 Alkyne is a selective STING (Stimulator of interferon genes) inhibitor with potential for research in autoimmune diseases, including systemic lupus erythematosus, scleroderma, and autoinflammatory diseases.
  • Selective STING inhibitor
  • Promising for autoimmune disease research
  • Supplied as a solid
  • Light yellow to brown appearance
  • High purity of 95.08%