Learn More
Medchemexpress LLC H-151 alkyne (compound H-151-AL) | 2244892-03-1 | 95.1% | 275.30 g/mol | C17H13N3O | 10 MG
SDP
Catalog No. 5000295286
Encompass_Preferred
H-151 Alkyne is an alkyne-labeled derivative of H-151 that functions as a selective STING (Stimulator of Interferon Genes) inhibitor for research use. It is provided as a purified small molecule intended for biochemical and cell-based assays to probe STING-mediated signaling and related autoinflammatory or autoimmune disease mechanisms.
  • Selective STING inhibition suitable for pathway studies.
  • Alkyne handle enables click chemistry labeling and probe conjugation.
  • Suitable for biochemical and cellular assays.
  • High purity (95.1%) for reproducible results.
  • Molecular weight 275.30 g/mol and formula C17H13N3O.
Catalog No. 50-002-95286 Supplier Medchemexpress LLC Supplier No. HY17656610MG
H-151 Alkyne is an alkyne-labeled derivative of H-151 that functions as a selective STING (Stimulator of Interferon Genes) inhibitor for research use. It is provided as a purified small molecule intended for biochemical and cell-based assays to probe STING-mediated signaling and related autoinflammatory or autoimmune disease mechanisms.
  • Selective STING inhibition suitable for pathway studies.
  • Alkyne handle enables click chemistry labeling and probe conjugation.
  • Suitable for biochemical and cellular assays.
  • High purity (95.1%) for reproducible results.
  • Molecular weight 275.30 g/mol and formula C17H13N3O.