Learn More
Bachem H-2-Mercapto-His-OH
SDP
Supplier:  Bachem 4014107.0001
Encompass_Preferred
H-2-Mercapto-His-OH, 2002-22-4, C6H9N3O2S, Mr 187.22, 1g. Key intermediate in the synthesis of the naturally occurring amino acid L-(+)-ergothioneine and analogs.
Catalog No. 50-259-221