Learn More
Bachem H-4,5-Dehydro-Leu-OH
SDP
Supplier:  Bachem 4018955.0001
Encompass_Preferred
H-4,5-Dehydro-Leu-OH, 87392-13-0, C6H11NO2, Mr 129.16, 1g. Precursor amino acid for tritium-labeled L-leucine. Specifically labeled bioactive peptides such as oxytocin can be conveniently synthesized by using this compound.
Catalog No. 50-259-189