Learn More
Bachem H-6-Diazo-5-oxo-Nle-OH
SDP
Supplier:  Bachem 4006579.0500
Encompass_Preferred
H-6-Diazo-5-oxo-Nle-OH, 157-03-9, C6H9N3O3, Mr 171.16, 500mg. L-DON, 6-Diazo-5-oxo-L-2-aminohexanoic acid, H-Glu(diazomethylketone)-OH. L-DON, which corresponds to Glu g-diazomethyl ketone, inactivates rat renal phosphate-dependent glutaminase and is active against various tumor systems. Irreversible inhibitor of g-glutamyl transferase (g-GT).
Catalog No. 50-259-141