Learn More
Enzo Life Sciences H-7 (50mg). CAS: 108930-17-2
SDP
Supplier:  Enzo Life Sciences BMLEI1480050
Encompass_Preferred
Inhibits PKA (Ki=3.0μM), PKG (Ki=5.8μM), MLCK (Ki=97μM) and PKC (Ki=6μM). Also inhibits myosin light chain kinase. Induces apoptotic DNA fragmentation and cell death. Inhibits telomerase activity in treated NPC-076 cells. Cell permeable. Alternative name: (+/-)-1-(5-Isoquinolinesulfonyl)-2-methylpiperazine . 2HCl. Purity: ≥98% (TLC). Solubility: Soluble in water (50mg/ml), 100% ethanol or DMSO. Long Term Storage: +4°C.
Catalog No. 50-201-0569