Learn More
Bachem Amyloid Beta Protein (1-42) (HFIP-treated)
SDP
Catalog No. 501946588
Encompass_Preferred
Amyloid Beta Protein (1-42) (HFIP-treated), H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH (HFIP-treated), 107761-42-2, C203H311N55O60S, Mr 4514.10, 1mg. Ab42 (HFIP-treated). The peptide was obtained by dissolving Amyloid b-Protein (1-42) in HFIP, aliquoting, and removing the solvent as described in the literature. The morphology of the resulting peptide was studied by transmission electron microscopy (TEM). The magenta arrow points to the nucleation centers where aggregation starts. The yellow arrow indicates the fibrils which are formed as long thin helical structures with regular twists.
Catalog No. 50-194-6588 Supplier Bachem Supplier No. 4090148.1000
Add to Cart
Add to Cart
Amyloid Beta Protein (1-42) (HFIP-treated), H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH (HFIP-treated), 107761-42-2, C203H311N55O60S, Mr 4514.10, 1mg. Ab42 (HFIP-treated). The peptide was obtained by dissolving Amyloid b-Protein (1-42) in HFIP, aliquoting, and removing the solvent as described in the literature. The morphology of the resulting peptide was studied by transmission electron microscopy (TEM). The magenta arrow points to the nucleation centers where aggregation starts. The yellow arrow indicates the fibrils which are formed as long thin helical structures with regular twists.