Learn More
AdipoGen H-8 . dihydrochloride (50 mg)
SDP
Supplier:  AdipoGen AGCR10001M050
Encompass_Preferred
Chemical. CAS: 113276-94-1. Formula: C12H15N3O2S . 2HCl. MW: 265.3 . 72.9. Potent cAMP- and cGMP-dependent protein kinase (PKA and PKG) inhibitor. Myosin light chain kinase (MLCK) inhibitor. Tool to study protein crystal structure-inhibitor interactions.
Catalog No. 50-205-3746