Learn More
Selleck Chemical LLC H 89 2HCl S1582-200mg
SDP
Catalog No. 503108491
Encompass_Preferred
H 89 2HCl is a potent PKA inhibitor with Ki of 48 nM in a cell-free assay 10-fold selective for PKA than PKG 500-fold greater selectivity than PKC MLCK calmodulin kinase II and casein kinase I/II H 89 2HCl induces autophagy
Catalog No. 50-310-8491 Supplier Selleck Chemical LLC Supplier No. S1582200MG
H 89 2HCl is a potent PKA inhibitor with Ki of 48 nM in a cell-free assay 10-fold selective for PKA than PKG 500-fold greater selectivity than PKC MLCK calmodulin kinase II and casein kinase I/II H 89 2HCl induces autophagy