Learn More
Enzo Life Sciences H-89 . 2HCl, (5mg), CAS Number: 127243-85-0
SDP
Supplier:  Enzo Life Sciences BMLEI1960005
Encompass_Preferred
Protein kinase inhibitor. Cell permeable potent and selective inhibitor of cAMP- and cGMP-dependent protein kinases (PKA, Ki=48nM, and PKG) and protein kinase Cµ (PKCµ). In contrast, most other protein kinase C (PKC) isotypes are much more weakly inhibited. Also inhibits Ca2+/calmodulin-dependent protein kinase II, casein kinase I and myosin light chain kinase. Induces apoptosis. Concentration range for cultured cells 30-100μM: osteoblastic cells, renal proximal tubule cells and Xenopus oocytes. . Alternative Name: N-[2-(p-Bromocinnamylamino)ethyl]-5-isoquinolinesulfonamide. 2HCl. Formula: C20H20BrN3O2S·2HCl. MW: 519.3. CAS Number: 127243-85-0. Purity: ≥99% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (25mg/ml), methanol and 50% ethanol/water (20mg/ml). Use/Stability: Store, as supplied, at 0-4°C for up to 1 year. Store solutions at -20°C for up to 4 months in DMSO. Long Term Storage: +4°C
Catalog No. NC9980061