Learn More
Medchemexpress LLC HY-15979A 50mg , H-89 (dihydrochloride) CAS:130964-39-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15979A50MG
Encompass_Preferred
HY-15979A 50mg H-89 (dihydrochloride) CAS: 130964-39-5 H-89 dihydrochloride is a potent and selective inhibitor of protein kinase A (PKA) with an IC50 of 48 nM and has weak inhibition on PKG, PKC, Casein Kinase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0182