Learn More
TARGETMOL CHEMICALS INC H-89 dihydrochloride 25MG
SDP
Catalog No. 502879997
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. H-89 dihydrochloride (5-Isoquinolinesulfonamide) is a potent inhibitor of protein kinase A (PKA; IC50 0.14 uM, Ki 48 nM). Purity 99.4%
Catalog No. 50-287-9997 Supplier TARGETMOL CHEMICALS INC Supplier No. T625025MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. H-89 dihydrochloride (5-Isoquinolinesulfonamide) is a potent inhibitor of protein kinase A (PKA; IC50 0.14 uM, Ki 48 nM). Purity 99.4%