Learn More
Medchemexpress LLC H-9 dihydrochloride | 116700-36-8 | MFCD00069286 | 99.0% | 324.23 g/mol | C11H15Cl2N3O2S | 25 MG
SDP
Catalog No. 5000211800
Encompass_Preferred
H-9 dihydrochloride is a small-molecule protein kinase inhibitor used in biochemical and cellular signaling research. It inhibits protein kinase A activity and reduces excitatory responses to serotonin, and is supplied as a light yellow solid suitable for preparation of DMSO stock solutions for in vitro assays.
  • Inhibits protein kinase A activity for signaling studies.
  • Reduces excitatory 5-HT responses in cellular assays.
  • Light yellow solid with high purity for research use.
  • Soluble in DMSO for convenient stock preparation.
  • Stable when stored sealed and kept away from moisture.
Catalog No. 50-002-11800 Supplier Medchemexpress LLC Supplier No. HY10092325MG
H-9 dihydrochloride is a small-molecule protein kinase inhibitor used in biochemical and cellular signaling research. It inhibits protein kinase A activity and reduces excitatory responses to serotonin, and is supplied as a light yellow solid suitable for preparation of DMSO stock solutions for in vitro assays.
  • Inhibits protein kinase A activity for signaling studies.
  • Reduces excitatory 5-HT responses in cellular assays.
  • Light yellow solid with high purity for research use.
  • Soluble in DMSO for convenient stock preparation.
  • Stable when stored sealed and kept away from moisture.