Learn More
Enzo Life Sciences H-9. HCl (50mg). CAS: 116970-50-4
SDP
Supplier:  Enzo Life Sciences BMLEI1950050
Encompass_Preferred
An excellent affinity ligand for chromatographic purification of protein kinases. Inhibits protein kinases (Ki's: PKA=1.9 µM; PKC=18 µM; PKG=0.87 µM; MLCK=70 µM). Alternative name: N-(2-Aminoethyl)-5-isoquinolinesulfonamide . HCl. Purity: ≥98% (TLC). Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-201-0602