Learn More
Bachem H-a-Difluoro-Me-DL-Orn-OH · HCl · H2O
SDP
Catalog No. 50259174
Encompass_Preferred
H-a-Difluoro-Me-DL-Orn-OH · HCl · H2O, 96020-91-6, C6H12F2N2O2 · HCl · H2O, Mr 236.65, 1g. Eflornithine · HCl · H2O, DFMO · HCl · H2O, a-Difluoromethylornithine · HCl · H2O. a-Difluoromethylornithine is a suicide inhibitor of ornithine decarboxylase. Initially developed for cancer treatment, eflornithine was found to be very efficient against trypanosomiasis. The ornithine derivative also found use in cosmetics, eflornithine-containing creams allow to reduce excessive hair growth (hirsutism). CAS Number (net): 67037-37-0, (hydrochloride): 68278-23-9.
Catalog No. 50-259-174 Supplier Bachem Supplier No. 4030547.1000
H-a-Difluoro-Me-DL-Orn-OH · HCl · H2O, 96020-91-6, C6H12F2N2O2 · HCl · H2O, Mr 236.65, 1g. Eflornithine · HCl · H2O, DFMO · HCl · H2O, a-Difluoromethylornithine · HCl · H2O. a-Difluoromethylornithine is a suicide inhibitor of ornithine decarboxylase. Initially developed for cancer treatment, eflornithine was found to be very efficient against trypanosomiasis. The ornithine derivative also found use in cosmetics, eflornithine-containing creams allow to reduce excessive hair growth (hirsutism). CAS Number (net): 67037-37-0, (hydrochloride): 68278-23-9.