Learn More
Haematologic Technologies Inc H ACTIVATED PROTEIN C 50UG
Supplier:  Haematologic Technologies Inc HCAPC0080
Encompass_Preferred
Coagulation Protein, Human activated Protein C, 50 micrograms, Native enzyme, MW=56,200, Mode of action: Anticoagulant, inactivates factors Va and VIIIa, Research use only, 50% (v/v) glycerol/Water, store at -20 Celsius.
Catalog No. 50-883-450