Learn More
Bachem H-Ala-Ala-Phe-AMC (free base)
SDP
Supplier:  Bachem 4002571.0050
Encompass_Preferred
H-Ala-Ala-Phe-AMC (free base), 62037-41-6, C25H28N4O5, Mr 464.52, 50mg. AAF-AMC (free base). AAF-AMC, fluorogenic substrate for tripeptidyl peptidases I and II and for tripeptide aminopeptidase EC 3.4.11.4.
Catalog No. 50-260-314