Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Ala-Hyp-OH | 76400-25-4 | 99.36% | 202.21 | 5 MG
SDP

Catalog No. 5000399626
Encompass_Preferred

H-Ala-Hyp-OH is a biologically active peptide identified by CAS number 76400-25-4, with a molecular weight of 202.21 and molecular formula C8H14N2O4. This peptide is provided as a white to off-white solid and is intended for research use only.

  • Biologically active peptide
  • Molecular formula C8H14N2O4
  • Molecular weight 202.21
  • High purity (99.36%)
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Supports various in vivo dissolution protocols

Catalog No. 50-003-99626 Supplier Medchemexpress LLC Supplier No. HYP53385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Ala-Hyp-OH is a biologically active peptide identified by CAS number 76400-25-4, with a molecular weight of 202.21 and molecular formula C8H14N2O4. This peptide is provided as a white to off-white solid and is intended for research use only.

  • Biologically active peptide
  • Molecular formula C8H14N2O4
  • Molecular weight 202.21
  • High purity (99.36%)
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Supports various in vivo dissolution protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.