Learn More
Medchemexpress LLC H-β-Ala-NH2·HCl-d4 | 1276197-35-3 | 128.59 | 1 MG
SDP
Catalog No. 5000460237
Encompass_Preferred
H-β-Ala-NH2·HCl-d4 is a deuterium-labeled compound, specifically a stable heavy isotope of H-β-Ala-NH2·HCl. Deuterium substitution has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only.
  • Deuterium labeled: Provides a stable heavy isotope for research applications.
  • Tracer compound: Can be used as a tracer in various studies.
  • Internal standard: Useful for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Potential for pharmacokinetic and metabolic studies: Deuteration may impact drug profiles, offering a tool for investigating these effects.
Catalog No. 50-004-60237 Supplier Medchemexpress LLC Supplier No. HYW007497S1MG
H-β-Ala-NH2·HCl-d4 is a deuterium-labeled compound, specifically a stable heavy isotope of H-β-Ala-NH2·HCl. Deuterium substitution has garnered attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only.
  • Deuterium labeled: Provides a stable heavy isotope for research applications.
  • Tracer compound: Can be used as a tracer in various studies.
  • Internal standard: Useful for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Potential for pharmacokinetic and metabolic studies: Deuteration may impact drug profiles, offering a tool for investigating these effects.