Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Ala-Pro-AFC | 125791-92-6 | MFCD00798773 | 99.4% | 397.35 g/mol | C18H18F3N3O4 | 10 MG
SDP

Catalog No. 5000281329
Encompass_Preferred

H-Ala-Pro-AFC is a fluorogenic dipeptide substrate for biochemical assays that measures dipeptidyl peptidase IV (DPP IV) and Xaa-Pro dipeptidase activity. It contains the AFC fluorophore (excitation/emission 380/500 nm) and is supplied as an off-white to light yellow solid or as DMSO solutions for easy reconstitution.

  • Fluorogenic substrate for dipeptidyl peptidase IV (DPP IV) and Xaa-Pro dipeptidase.
  • Excitation/emission at 380/500 nm.
  • High purity: 99.4% (LCMS).
  • Molecular weight 397.35 g/mol.
  • Supplied as lyophilized solid or DMSO solution for reconstitution.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-81329 Supplier Medchemexpress LLC Supplier No. HYW43647110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Ala-Pro-AFC is a fluorogenic dipeptide substrate for biochemical assays that measures dipeptidyl peptidase IV (DPP IV) and Xaa-Pro dipeptidase activity. It contains the AFC fluorophore (excitation/emission 380/500 nm) and is supplied as an off-white to light yellow solid or as DMSO solutions for easy reconstitution.

  • Fluorogenic substrate for dipeptidyl peptidase IV (DPP IV) and Xaa-Pro dipeptidase.
  • Excitation/emission at 380/500 nm.
  • High purity: 99.4% (LCMS).
  • Molecular weight 397.35 g/mol.
  • Supplied as lyophilized solid or DMSO solution for reconstitution.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.