Learn More
Carbosynth LLC. H-Ala-Tyr-Pro-Gly-Lys-Phe-NH2, 5mg, MW: 680.809999999999, Molecular formula: C34H48N8O7, CAS# 352017-71-1
SDP
Supplier:  Carbosynth LLC. PAR3674PI5MG
Encompass_Preferred
Synonyms: H-Ala-Tyr-Pro-Gly-Lys-Phe-NH2, AYPGKF-Amide, Application: Selective Protease-Activated Receptor 4 (PAR4) Agonist, Salt Form:, Acetate Salt, Storage Temperature: -20°C, References: M.D. Hollenberg, M. Saifeddine, B. Al-Ani, and Y. Gui, Can. J. Physiol. Pharmacol., 77, 458 (1999)., T.R. Faruqi, E.J. Weiss, M.J. Shapiro, W. Huang, and S.R. Coughlin, J. Biol. Chem., 275, 19728 (2000)., M.D. Hollenberg, M. Saifeddine, S. Sandhu, S. Houle, and N. Vergnolle, Br. J. Pharmacol., 143, 443 (2004).
Catalog No. 50-168-6183