Learn More
Medchemexpress LLC H-allo-Thr(tBu)-OH | 201353-89-1 | 97.0% | 175.23 g/mol | C8H17NO3 | 1 G
SDP
Catalog No. 5000209957
Encompass_Preferred
H-allo-Thr(tBu)-OH is a tert-butyl protected allo-threonine derivative supplied for research use. It is used as a protected amino acid building block in peptide synthesis and biochemical studies.
  • Tert-butyl protected allo-threonine suitable for peptide synthesis.
  • Provided with supporting documents such as SDS, COA, and spectral data.
  • Molecular formula C8H17NO3 and molecular weight 175.23 g/mol.
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-09957 Supplier Medchemexpress LLC Supplier No. HYW0513511G
H-allo-Thr(tBu)-OH is a tert-butyl protected allo-threonine derivative supplied for research use. It is used as a protected amino acid building block in peptide synthesis and biochemical studies.
  • Tert-butyl protected allo-threonine suitable for peptide synthesis.
  • Provided with supporting documents such as SDS, COA, and spectral data.
  • Molecular formula C8H17NO3 and molecular weight 175.23 g/mol.
  • Intended for research use only; not for human consumption.