Learn More
Haematologic Technologies Inc H ALPHA-2 ANTIPLASMIN 100UG
Catalog No. 50883476
Encompass_Preferred
Inhibitor, Human alpha-2 Antiplasmin (Alpha-2 anti-plasmin), 100 micrograms, Purified from plasma, Action: Prevents the binding of plasmin to fibrin, Research use only, Supplied in phosphate buffer, store at -80 Celsius.
Catalog No. 50-883-476 Supplier Haematologic Technologies Inc Supplier No. HA2AP0230
Add to Cart
Add to Cart
Inhibitor, Human alpha-2 Antiplasmin (Alpha-2 anti-plasmin), 100 micrograms, Purified from plasma, Action: Prevents the binding of plasmin to fibrin, Research use only, Supplied in phosphate buffer, store at -80 Celsius.