Learn More
Electron Microscopy Sciences H And E Slide Stainer
SDP
Catalog No. 5033046
Encompass_Preferred
The new EMS slide stainer offers the end user the ability to have a stainer with 30 stations and a continuous load option with multiple protocols. This state of the art stainer which is only 32" long has a built in fume extraction as well as the ability to connect to a lab fume hood.
Catalog No. 50-330-46 Supplier Electron Microscopy Sciences Supplier No. 63122
Add to Cart
Add to Cart
Request a Quote
The new EMS slide stainer offers the end user the ability to have a stainer with 30 stations and a continuous load option with multiple protocols. This state of the art stainer which is only 32" long has a built in fume extraction as well as the ability to connect to a lab fume hood.