Learn More
Medchemexpress LLC HY-112688 10mg Medchemexpress, H-Arg-4MβNA CAS:60285-94-1 Purity:>98%
SDP
Catalog No. 501969141
Encompass_Preferred
Medchemexpress, HY-112688 10mg H-Arg-4MβNA CAS:60285-94-1 H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9141 Supplier Medchemexpress LLC Supplier No. HY11268810MG
Medchemexpress, HY-112688 10mg H-Arg-4MβNA CAS:60285-94-1 H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.