Learn More
Medchemexpress LLC H-Arg-4MβNA | 60285-94-1 | 99.6% | 329.40 | 50 MG
SDP
Catalog No. 5000436996
Encompass_Preferred
H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis.
  • Used for the detection of enzyme activity in gel electrophoresis.
Catalog No. 50-004-36996 Supplier Medchemexpress LLC Supplier No. HY11268850MG
H-Arg-4MβNA is a substrate for cathepsin H, used for the detection of enzyme activity in gel electrophoresis.
  • Used for the detection of enzyme activity in gel electrophoresis.