Learn More
Medchemexpress LLC H-arg-4mbna | 60285-94-1 | MFCD00058039 | 99.6% | 329.40 g/mol | C17H23N5O2 | 5 MG
SDP
Catalog No. 5000262459
Encompass_Preferred
H-Arg-4MβNA is a synthetic peptide substrate for cathepsin H used to detect enzymatic activity in gel electrophoresis. Proteolytic cleavage releases a fluorogenic 4-methylumbelliferone derivative, enabling sensitive visualization and quantitation of cathepsin H activity. Supplied as a solid or as a ready-to-use 10 mM solution for research applications.
  • Substrate for cathepsin H suitable for gel electrophoresis assays.
  • Releases a fluorogenic 4-methylumbelliferone derivative upon cleavage.
  • High purity (99.62%) for reliable experimental results.
  • Offered as solid and as a 10 mM solution in water.
  • Available in multiple pack sizes to suit assay scale.
  • Includes supporting documentation such as COA, SDS, and LC-MS.
Catalog No. 50-002-62459 Supplier Medchemexpress LLC Supplier No. HY1126885MG
H-Arg-4MβNA is a synthetic peptide substrate for cathepsin H used to detect enzymatic activity in gel electrophoresis. Proteolytic cleavage releases a fluorogenic 4-methylumbelliferone derivative, enabling sensitive visualization and quantitation of cathepsin H activity. Supplied as a solid or as a ready-to-use 10 mM solution for research applications.
  • Substrate for cathepsin H suitable for gel electrophoresis assays.
  • Releases a fluorogenic 4-methylumbelliferone derivative upon cleavage.
  • High purity (99.62%) for reliable experimental results.
  • Offered as solid and as a 10 mM solution in water.
  • Available in multiple pack sizes to suit assay scale.
  • Includes supporting documentation such as COA, SDS, and LC-MS.