Learn More
Bachem H-Arg-Ala-OH
SDP
Supplier:  Bachem 4017629.1000
Encompass_Preferred
H-Arg-Ala-OH Acetate, 40968-45-4, C9H19N5O3, Mr 245.28, 1g. A salt taste-enhancing dipeptide. Arg-Ala inhibits angiotensin 1-converting enzyme (ACE) and yeast bleomycin hydrolase.
Catalog No. 50-259-432